Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNAH12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557974
Description
Immunogen sequence: IHGLYLDGAR WDRESGLLAE QYPKLLFDLM PIIWIKPTQK SRIIKSDAYV CP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
DNAH12 gene ontology annotations related to this gene include microtubule-based movement.
Specifications
DNAH12 | |
Polyclonal | |
Unconjugated | |
DNAH12 | |
4921531P07Rik; axonemal beta dynein heavy chain 12; Axonemal dynein heavy chain 12-like protein; Axonemal dynein heavy chain 7-like protein; axonemal dynein heavy chain isotype3; Bm259; ciliary dynein heavy chain 12; DHC3; DLP12; Dnah12; DNAH12L; DNAH7L; DNAHC12; Dnahc3; Dnahc7c; Dnahc7l; DNHD2; dynein axonemal heavy chain 12; dynein heavy chain 12, axonemal; dynein heavy chain 12, axonemal-like; Dynein heavy chain 7-like, axonemal; dynein heavy chain domain 2; dynein heavy chain domain-containing protein 2; dynein, axonemal, heavy chain 12; dynein, axonemal, heavy polypeptide 12; dynein, heavy chain-5; Gm284; Gm74; Gm907; Hdhc3; HL19; HL-19 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
201625 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZR08 | |
DNAH12 | |
Recombinant protein corresponding to Human DNAH12. Recombinant protein control fragment (Product #RP-96418). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction