Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAH2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548744

Catalog No. PA548744

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules. Dynein has ATPase activity; the force-producing power stroke is thought to occur on release of ADP. Involved in sperm motility; implicated in sperm flagellar assembly.
Specifications

Specifications

DNAH2
Polyclonal
PBS with 0.09% sodium azide
Q9P225
Dynein heavy chain 2, axonemal, Axonemal beta dynein heavy chain 2, Ciliary dynein heavy chain 2, Dynein heavy chain domain-containing protein 3, DNAH2, DNAHC2, DNHD3, KIAA1503
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
146754
Western Blot
Unconjugated
DNAH2
Liquid
DNAH2
KLH conjugated synthetic peptide between 440-468 amino acids from the N-terminal region of human DNAH2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit