Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAH6 Monoclonal Antibody (OTI5C2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812184


Description
DNAH6 Monoclonal antibody specifically detects DNAH6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
DNAH6 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q10588 | |
Mouse | |
Protein A/G | |
RUO | |
683 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5C2 | |
Unconjugated | |
Bst1 | |
9830168K20; A730004I20Rik; axo a; Axonemal beta dynein heavy chain 6; axonemal dynein heavy chain; ciliary dynein heavy chain 6; DNAH6; Dnahc6; DNHL1; dynein axonemal heavy chain 6; dynein heavy chain 6, axonemal; dynein heavy chain-like 1; dynein, axon, heavy chain 6; dynein, axonemal, heavy chain 6; dynein, axonemal, heavy polypeptide 6; HL2; HL-2; KIAA1697; mdhc6; mKIAA1697 | |
Human recombinant protein fragment corresponding to amino acids 29-318 of human BST1 (NP_004325) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction