Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJA2 Mouse anti-Human, Rat, Canine, Monkey, PE-Cyanine7, Clone: OTI3A10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271565PECY7

Catalog No. N271565PEC7


Add to cart

Description

Description

DNAJA2 Monoclonal antibody specifically detects DNAJA2 in Human, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

DNAJA2
OTI3A10
Flow Cytometry
Affinity Purified, Conjugated
DNAJA2
Full length recombinant protein of human DNAJA2 (NP_005871) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication
Primary
10294.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
cell cycle progression 3 protein, Cell cycle progression restoration gene 3 protein, CPR3PRO3015, Dj3, DJA2, DNAJ, DnaJ (Hsp40) homolog, subfamily A, member 2, dnaJ homolog subfamily A member 2, DNJ3, HIRA interacting protein 4, HIRA-interacting protein 4, HIRIP4RDJ2, Renal carcinoma antigen NY-REN-14
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit