Learn More
DNAJA2 Mouse anti-Canine, Human, Non-human primate, Rat, Clone: OTI3A10, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525232
Catalog No. PIMA525232
Add to cart
Description
Description
The protein encoded by this gene belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain. The product of this gene works as a cochaperone of Hsp70s in protein folding and mitochondrial protein import in vitro.
Specifications
Specifications
DNAJA2
Monoclonal
0.37 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O35824, O60884
CPR3, DJ3, DJA2, DNAJ, DNJ3, HIRIP4, PRO3015, RDJ2, HIRA interacting protein 4, HIRA-interacting protein 4, cell cycle progression 3 protein, cell cycle progression restoration gene 3 protein, dnaJ homolog subfamily A member 2, renal carcinoma antigen NY-REN-14
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10294, 478143, 716353, 84026
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI3A10
Unconjugated
DNAJA2
Liquid
Dnaja2
Full length human recombit protein of DNAJA2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate, Rat