Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJB1 Mouse anti-Human, Clone: OTI1B12, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502245

Catalog No. 50-167-3319


Add to cart

Description

Description

Interacts with HSP70 and can stimulate its ATPase activity. Stimulates the association between HSC70 and HIP.
Specifications

Specifications

DNAJB1
Monoclonal
0.38 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P25685
HSPF1, Hdj1, Hsp40, RSPH16B, Sis1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3337
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1B12
Unconjugated
DNAJB1
Liquid
DNAJB1
Full length human recombit protein of human DNAJB1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit