Learn More
Aviva Systems Biology Corp DNAJB5 Antibody - N-terminal region (AVARP09012_T100) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP09012_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DNAJB5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DNAJB5. Alternate names: Hsc40ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62564