Learn More
Invitrogen™ DNAJC11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144019
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human SH-SY5Y whole cell, human A549 whole cell, rat liver tissue, rat skeletal muscle tissue, mouse liver tissue, mouse skeletal muscle tissue, human HepG2 whole cell. ICC/IF: Hela cell. Flow: PC-3 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Belongs to the DNAJC11 family. Three named isoforms produced by alternative splicing.
Specifications
DNAJC11 | |
Polyclonal | |
Unconjugated | |
Dnajc11 | |
dJ126A5.1; DnaJ (Hsp40) homolog, subfamily C, member 11; DnaJ heat shock protein family (Hsp40) member C11; dnaJ homolog subfamily C member 11; DNAJC11; E030019A03Rik; novel DnaJ domain-containing protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230935, 362666, 55735 | |
-20°C | |
Lyophilized |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
Q5U458, Q9NVH1 | |
Dnajc11 | |
E.coli-derived human DNAJC11 recombinant protein (Position: N12-Q551). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.