Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC14 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548516

Catalog No. PA548516

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Regulates the export of target proteins, such as DRD1, from the endoplasmic reticulum to the cell surface.
Specifications

Specifications

DNAJC14
Polyclonal
PBS with 0.09% sodium azide
Q6Y2X3
DnaJ homolog subfamily C member 14, DnaJ protein homolog 3, Dopamine receptor-interacting protein of 78 kDa, DRIP78, Human DnaJ protein 3, hDj-3, DNAJC14, DRIP78, HDJ3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
85406
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DNAJC14
Liquid
DNAJC14
KLH conjugated synthetic peptide between 184-213 amino acids from the Central region of human DNAJC14.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit