Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC15 Polyclonal Antibody

DNAJC15 Polyclonal Antibody

Supplier:  Thermo Scientific PA550418

Catalog No. PA550418


Only null left
Add to Cart

Description

Description

DNAJC15 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNAJC15
Polyclonal
Unconjugated
DNAJC15
1110003P16Rik; cell growth-inhibiting gene 22 protein; DnaJ (Hsp40) homolog, subfamily C, member 15; DnaJ (Hsp40) homolog, subfamily D, member 1; DNAJ domain-containing; DnaJ heat shock protein family (Hsp40) member C15; dnaJ homolog subfamily C member 15; DNAJC15; Dnajd1; GIG22; HSD18; MCJ; methylation-controlled J protein
Rabbit
Antigen affinity chromatography
RUO
29103
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.5 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9Y5T4
DNAJC15
Fusion protein corresponding to a region derived from internal residues of human DnaJ (Hsp40) homolog, subfamily C, member 15.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.