Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC15 Rabbit anti-Human, Clone: SY26-04, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267439

Catalog No. NBP267439


Add to cart

Description

Description

DNAJC15 Monoclonal antibody specifically detects DNAJC15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DNAJC15
SY26-04
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
Cell growth-inhibiting gene 22 protein, DnaJ (Hsp40) homolog, subfamily C, member 15, DnaJ (Hsp40) homolog, subfamily D, member 1, DNAJ domain-containing, dnaJ homolog subfamily C member 15, DNAJD1, MCJHSD18, Methylation-controlled J protein
Rabbit
IgG
100μL
Primary
29103
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Purified
DNAJC15
Recombinant protein
Protein A purified
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit