Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC24 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548741

Catalog No. PA548741

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Stimulates the ATPase activity of several Hsp70-type chaperones. This ability is enhanced by iron-binding. The iron- bound form is redox-active and can function as electron carrier. Plays a role in the diphthamide biosynthesis, a post-translational modification of histidine which occurs in translation elongation factor 2 (EEF2) which can be ADP-ribosylated by diphtheria toxin and by Pseudomonas exotoxin A (Eta).
Specifications

Specifications

DNAJC24
Polyclonal
PBS with 0.09% sodium azide
Q6P3W2
DnaJ homolog subfamily C member 24, CSL-type zinc finger-containing protein 3, Diphthamide biosynthesis protein 4, DNAJC24, DPH4, ZCSL3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
120526
Western Blot
Unconjugated
DNAJC24
Liquid
DNAJC24
KLH conjugated synthetic peptide between 53-81 amino acids from the Central region of human DNAJC24.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit