Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC27 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548442

Catalog No. PA548442

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GTPase which can activate the MEK/ERK pathway and induce cell transformation when overexpressed. May act as a nuclear scaffold for MAPK1, probably by association with MAPK1 nuclear export signal leading to enhanced ERK1/ERK2 signaling.
Specifications

Specifications

DNAJC27
Polyclonal
PBS with 0.09% sodium azide
Q9NZQ0
DnaJ homolog subfamily C member 27, Rab and DnaJ domain-containing protein, DNAJC27, RABJS, RBJ
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51277
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DNAJC27
Liquid
DNAJC27
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human DNAJC27.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit