Learn More
Abnova™ DNAJC5B 293T Cell Transient Overexpression Lysate (Denatured) (T03)
Human overexpression lysate
Manufacturer:  Abnova™ H00085479T03
Catalog No. 89-942-213
Add to cart
Specifications
Specifications
Western Blot
293T
NM_033105
pCMV-DNAJC5B full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
DNAJC5B 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 22kDa
Transient overexpression cell lysate was tested with anti-DNAJC5B antibody (Mfr. No. H00085479-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)