Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJC5B Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2848080.1ML
Description
DNAJC5B Polyclonal specifically detects DNAJC5B in Mouse samples. It is validated for Western Blot.Specifications
DNAJC5B | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
85479 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Beta cysteine string protein, Beta-CSP, CSP-beta, DnaJ (Hsp40) homolog, subfamily C, member 5 beta, dnaJ homolog subfamily C member 5B, dnaJ homolog subfamily C member X, MGC26226 | |
The immunogen is a synthetic peptide directed towards the middle terminal region of mouse DNAJC5B | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction