Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNAJC9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557199
Description
Immunogen sequence: EPRIRNIIQQ AIDAGEVPSY NAFVKESKQK MNARKRRAQE EAKEAEMSRK ELGLDEGVDS LKAAIQSRQK DRQKEMDNFL AQMEAKYCKS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 91%.
DjC9 (DnaJ subfamily C member 9) belongs to the Hsp40 (heat shock protein 40) family of proteins that function as co-chaperones to Hsp70.Hsp40 proteins are classified into 3 main subfamilies (A, B, and C). The families are defined by the presence of particular domains which include a highly conserved alpha-helical N-terminal domain termed the J-domain, a glycine/phenylalanine-rich region, a cysteine-rich region, and a C-terminal beta-sheet containing domain. DjC9 is part of subfamily C that bears only the J-domain. DjC9 has been shown to interact with HSP70 through its J domain and activate its ATPase activity.
Specifications
DNAJC9 | |
Polyclonal | |
Unconjugated | |
DNAJC9 | |
5330419I01Rik; AU020082; DnaJ (Hsp40) homolog, subfamily C, member 9; DnaJ heat shock protein family (Hsp40) member C9; dnaJ homolog subfamily C member 9; DnaJ protein SB73; Dnajc9; DnaJ-like factor; HDJC9; JDD1; SB73 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23234 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WXX5 | |
DNAJC9 | |
Recombinant protein corresponding to Human DNAJC9. Recombinant protein control fragment (Product #RP-95737). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction