Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAL1 Mouse anti-Human, Alexa Fluor™ 532, Clone: 290000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242657AF532

Catalog No. NP24267A32


Add to cart

Description

Description

DNAL1 Monoclonal antibody specifically detects DNAL1 in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

DNAL1
290000
Western Blot, ELISA
C14orf168MGC12435,1700010H15RiK, chromosome 14 open reading frame 168, dynein light chain 1, axonemal, dynein, axonemal, light chain 1
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
DNAL1
Recombinant human DNAL1 (1-190aa) purified from E. coli
0.1 mL
Primary
83544
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit