Learn More
DNALI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 176011AP
Catalog No. 50-173-1678
Add to cart
Description
Description
This gene is the human homolog of the Chlamydomonas inner dynein arm gene, p28. The precise function of this gene is not known, however, it is a potential candidate for immotile cilia syndrome (ICS). Ultrastructural defects of the inner dynein arms are seen in patients with ICS. Immotile mutant strains of Chlamydomonas, a biflagellated algae, exhibit similar defects.
Specifications
Specifications
DNALI1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14645, Q4FZV3, Q8BVN8
DNALI1, hp28, P28
Rabbit
IgG
150 μL
-20°C
Primary
298524, 75563, 7802
Immunoprecipitation, Western Blot
Unconjugated
DNALI1
Liquid
Dnali1
DNALI1 Fusion Protein Ag11652
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat