Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DNase I from Bovine Pancreas

Derived from Bovine pancreas.

$89.70 - $151.00

Specifications

Source Material Bovine pancreas
Format Lyophilized powder
Product Type DNase I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ14340MA View Documents Thermo Scientific™
J14340MA
10mg Each for $89.70
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ14340FPL View Documents Affymetrix/USB™
J14340FPL
20mg Each for $151.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • Synonym: E.C.3.1.21.1
  • MDL: MFCD00130917
Specifications

Specifications

Bovine pancreas
Lyophilized powder
DNase I
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit