Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DNase I, RNase-free: PROMO

Endonuclease that nonspecifically cleaves single- and double-stranded DNA to release di-, tri-, and oligonucleotides with 5'-phosphate and 3'-OH groups.

Manufacturer:  Thermo Scientific™ EN0521

 View more versions of this product

Catalog No. FEREN0521PM

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Highlights

  • Recombinant enzyme
  • Purified from non-animal host with a lower level of intrinsic RNases

Applications

  • Preparation of DNA-free RNA
  • Removal of template DNA followingin vitrotranscription
  • Preparation of DNA-free RNA prior to RT-PCR and RT-qPCR
  • DNA labeling by nick-translation in conjunction with DNA Polymerase I
  • Studies of DNA-protein interactions by DNase I, RNase-free footprinting
  • Generation of a library of randomly overlapping DNA inserts. Reaction buffer containing Mn2+ is used

Note

DNase I is sensitive to physical denaturation. Mix gently by inverting the tube. Do not vortex.

Specifications

Specifications

1U/μL
E.coli cells with a cloned gene encoding bovine DNase
1000U
metal chelators, transition metals (e.g., Zn) in millimolar concentrations, SDS (even at concentrations less than 0.1%), reducing agents (DTT and beta-mercaptoethanol), ionic strength above 50-100 mM
29 kDa monomer
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit