Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNASE1 Rabbit anti-Human, DyLight 550, Clone: 6, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290240R

Catalog No. NB038248


Add to cart

Description

Description

DNASE1 Monoclonal antibody specifically detects DNASE1 in Human samples. It is validated for ELISA, Immunofluorescence, Immunofluorescence
Specifications

Specifications

DNASE1
6
ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence
Purified
Rabbit
IgG
0.1 mL
Apoptosis, DNA Repair, DNA replication Transcription Translation and Splicing
Primary
1773
ELISA, Immunofluorescence
DyLight 550
50mM Sodium Borate
deoxyribonuclease IFLJ38093, deoxyribonuclease-1, DKFZp686H0155, DNase I, DNase I, lysosomal, DNL1FLJ44902, Dornase alfa, DRNI, EC 3.1.21.1, human urine deoxyribonuclease I
Recombinant Human DNASE1 Protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit