Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNASE1L1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549091

Catalog No. PA549091

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a deoxyribonuclease protein that shows high sequence similarity to DNase I. The encoded protein is localized to the endoplasmic reticulum and modified by N-linked glycosylation. Alternate transcriptional splice variants encoding the same protein have been observed. [provided by RefSeq, Jan 2015]
Specifications

Specifications

DNASE1L1
Polyclonal
PBS with 0.09% sodium azide
P49184
Deoxyribonuclease-1-like 1, 3121-, DNase X, Deoxyribonuclease I-like 1, DNase I-like 1, Muscle-specific DNase I-like, XIB, DNASE1L1, DNAS1L1, DNL1L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1774
Western Blot
Unconjugated
DNASE1L1
Liquid
DNASE1L1
KLH conjugated synthetic peptide between 177-204 amino acids from the Central region of human DNASE1L1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit