Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAzol™ Reagent

$134.90 - $458.00

Specifications

Format Liquid
For Use With (Application) Complete and ready to use reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin
Color Blue-green
Product Type DNA Extraction Reagent
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ60637AH View Documents Alfa Aesar™
J60637AH
200mL Each for $458.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ60637AD View Documents Alfa Aesar™
J60637AD
50mL Each for $134.90
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ60637AE View Documents Alfa Aesar™
J60637AE
100mL Each for $246.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • Complete and ready to use reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin
Specifications

Specifications

Liquid
Complete and ready to use reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin
Blue-green
DNA Extraction Reagent
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit