Learn More
DNM1L Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI4F6, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA506153
Catalog No. 50-167-6498
Add to cart
Description
Description
A human dynamin-related protein, DRP1 contributes to mitochondrial division in mammalian cells. It plays this important role in mitochondrial fission at steady state and during apoptosis. DRP1 is required for proper cellular distribution of mitochondria, and in mutant neurons, mitochondria are largely absent from synapses, thus providing a genetic tool to assess the role of mitochondria at synapses.
Specifications
Specifications
DNM1L
Monoclonal
Unconjugated
DNM1L
Liquid
Dnm1l
Full length human recombit protein of human DNM1L produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
OTI4F6
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00429, O35303, Q8K1M6
DLP1, DRP1, DVLP, DYMPLE, EMPF, EMPF1, HDYNIV, OPA5
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10059, 114114, 477649, 74006