Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNM1L Mouse anti-Human, Mouse, Rat, Clone: OTI2E7, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA506115S
Description
A human dynamin-related protein, DRP1 contributes to mitochondrial division in mammalian cells. It plays this important role in mitochondrial fission at steady state and during apoptosis. DRP1 is required for proper cellular distribution of mitochondria, and in mutant neurons, mitochondria are largely absent from synapses, thus providing a genetic tool to assess the role of mitochondria at synapses.Specifications
DNM1L | |
Monoclonal | |
Unconjugated | |
DNM1L | |
DLP1, DRP1, DVLP, DYMPLE, EMPF, EMPF1, HDYNIV, OPA5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10059, 114114, 74006 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2E7 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O00429, O35303, Q8K1M6 | |
Dnm1l | |
Full length human recombit protein of human DNM1L produced in HEK293T cell. | |
30 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction