Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNM1L Monoclonal Antibody (OTI3F3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526250
Description
DNM1L Monoclonal antibody specifically detects DNM1L in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
DNM1L | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00429 | |
DNM1L | |
Full length human recombinant protein of DNM1L produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3F3 | |
Unconjugated | |
DNM1L | |
6330417M19Rik; AI450666; C-terminal region; Dlp1; dnm1l; Dnm1p/Vps1p-like protein; Dnml1; Dnmlp1; Drp1; DVLP; DYMPLE; dynamin 1 like; dynamin 1-like; dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-1-like protein-like protein; Dynamin-like protein; dynamin-like protein 1; dynamin-like protein 4; Dynamin-like protein IV; dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; EGK_03514; EMPF; FLJ41912; HDYNIV; N-terminal region; python; similar to dynamin 1-like; VPS1; zgc:66163 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10059 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction