Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNM2 Mouse anti-Human, Clone:3F5C7, Abnova™

Mouse monoclonal antibody raised against recombinant human DNM2.

Manufacturer:  Abnova Corporation MAB17595

Catalog No. 89-334-019


Add to cart

Description

Description

Dynamins represent one of the subfamilies of GTP-binding proteins. These proteins share considerable sequence similarity over the N-terminal portion of the molecule, which contains the GTPase domain. Dynamins are associated with microtubules. They have been implicated in cell processes such as endocytosis and cell motility, and in alterations of the membrane that accompany certain activities such as bone resorption by osteoclasts. Dynamins bind many proteins that bind actin and other cytoskeletal proteins. Dynamins can also self-assemble, a process that stimulates GTPase activity. Four alternatively spliced transcripts encoding different proteins have been described. Additional alternatively spliced transcripts may exist, but their full-length nature has not been determined. [provided by RefSeq]
Specifications

Specifications

DNM2
3F5C7
dynamin 2
In PBS (0.05% sodium azide)
Liquid
Mouse
IgG2a
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry
Immunohistochemistry
The optimal working dilution should be determined by the end user.
DNM2
CMTDI1, CMTDIB, DI-CMTB, DYN2, DYNII
Recombinant protein corresponding to amino acid 520-744 of human DNM2 from E. coli.
100μg
Antibody
1785
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit