Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1 Mouse anti-Human, Clone: DNMT1/2061, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen DNMT1
Clone DNMT1/2061
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122816
View Documents
Novus Biologicals
NBP30767120UG
20 μg
Each for $138.37
Add to cart
 
NB122815
View Documents
Novus Biologicals
NBP30767110UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

DNMT1 Monoclonal antibody specifically detects DNMT1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

DNMT1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Recombinant human DNMT1 protein fragment (around aa 767-912) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
DNMT1/2061
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
10 mM PBS with 0.05% BSA
1786
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit