Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNMT1 Antibody (4G11-C7), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259344G
Description
DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse, Fish, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DNMT1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DNMT1 | |
Raised against a partial protein corresponding to amino acids 620-950 of human DNMT1 | |
100 μL | |
Primary | |
Detects 180kDa. It will cross-react with mouse DNMT1. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry | |
4G11-C7 | |
Western Blot, Immunohistochemistry | |
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992 | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer, Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators | |
1786 | |
Human, Mouse, Fish, Zebrafish | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction