Learn More
DNMT1 Mouse anti-Human, Mouse, Fish, Zebrafish, DyLight 680, Clone: 4G11-C7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259344FR
Catalog No. NBP259344FR
Add to cart
Description
Description
DNMT1 Monoclonal antibody specifically detects DNMT1 in Human, Mouse, Fish, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
DNMT1
4G11-C7
Western Blot, Immunohistochemistry
Purified
DNMT1
Raised against a partial protein corresponding to amino acids 620-950 of human DNMT1
Protein G purified
Angiogenesis, Cancer, Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
Primary
1786
Human, Mouse, Fish, Zebrafish
Western Blot, Immunohistochemistry
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Mouse
IgG1 Kappa
100 ul
Store at 4C in the dark.
Monoclonal
Detects 180kDa. It will cross-react with mouse DNMT1.