Learn More
DNMT1 Mouse anti-Human, Primate, Alexa Fluor 700, Clone: 7C11A2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237335AF700
Catalog No. NBP237335X
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
DNMT1
7C11A2
Western Blot, ELISA, Immunohistochemistry
Purified
DNMT1
Purified recombinant fragment of human DNMT1 (AA: 1448-1594) expressed in E. Coli.
Protein A or G purified
Angiogenesis, Cancer, Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
Primary
1786
ELISA, Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Primate