Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT1 Mouse anti-Human, Primate, FITC, Clone: 7C11A2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237335F

Catalog No. NBP237335F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

DNMT1
7C11A2
ELISA, Immunohistochemistry
Purified
DNMT1
Purified recombinant fragment of human DNMT1 (AA: 1448-1594) expressed in E. Coli.
Protein A or G purified
Angiogenesis, Cancer, Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
Primary
1786
ELISA, Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit