Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP153132

 View more versions of this product

Catalog No. NBP153132

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Dnmt2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Dnmt2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
TRDMT1
Synthetic peptides corresponding to TRDMT1(tRNA aspartic acid methyltransferase 1) The peptide sequence was selected from the N terminal of TRDMT1. Peptide sequence MILMSPPCQPFTRIGRQGDMTDSRTNSFLHILDILPRLQKLPKYILLENV.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Chicken: 92%; Xenopus: 85%; Canine: 85%.
Human
Western Blot
Western Blot 1:100-1:2000
O14717
DMNT2, DNA (cytosine-5-)-methyltransferase 2, DNA (cytosine-5)-methyltransferase-like protein 2, DNA methyltransferase homolog HsaIIP, DNA methyltransferase-2, DNA MTase homolog HsaIIP, Dnmt2, EC 2.1.1, EC 2.1.1.29, M.HsaIIP, MHSAIIP, PuMet, RNMT1, tRNA (cytosine-5-)-methyltransferase, tRNA aspartic acid methyltransferase 1
Rabbit
IgG
100 ul
Chromatin Modifiers, Chromatin Research, Epigenetics
Primary
1787
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit