Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNMT3A Mouse anti-Human, Clone: PCRP-DNMT3A-1E2, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | DNMT3A |
---|---|
Clone | PCRP-DNMT3A-1E2 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122818
|
Novus Biologicals
NBP30767220UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
DNMT3A Monoclonal specifically detects DNMT3A in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
DNMT3A | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cancer, Chromatin Research, Epigenetics, Innate Immunity, Tumor Suppressors | |
10 mM PBS with 0.05% BSA | |
1788 | |
IgG1 κ | |
Protein A or G purified |
PCRP-DNMT3A-1E2 | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA Methyltransferase 3 Alpha, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA, TBRS | |
Recombinant human full-length DNMT3A protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title