Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3A Mouse anti-Human, Clone: PCRP-DNMT3A-1E2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30767210UG

 View more versions of this product

Catalog No. NB122817


Add to cart

Description

Description

DNMT3A Monoclonal specifically detects DNMT3A in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

DNMT3A
PCRP-DNMT3A-1E2
Unconjugated
10 mM PBS with 0.05% BSA
DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA Methyltransferase 3 Alpha, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA, TBRS
Recombinant human full-length DNMT3A protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Western Blot, Immunohistochemistry (Paraffin), Peptide Array
0.2 mg/ml
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Purified
Mouse
IgG1 Kappa
100 μg
Adaptive Immunity, Cancer, Chromatin Research, Epigenetics, Innate Immunity, Tumor Suppressors
Primary
1788
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit