Learn More
DNMT3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 193661AP
Catalog No. 50-173-2390
Add to cart
Description
Description
TREF1 encodes a zinc-finger transcriptional regulating protein which interacts with CBP/p300 to regulate the human gene CYP11A1.
Specifications
Specifications
DNMT3A
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88508, Q1LZ53, Q9Y6K1
DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, DNMT3A, DNMT3A2, M.HsaIIIA
Rabbit
IgG
150 μL
-20°C
Primary
13435, 1788, 444984
Immunohistochemistry (Paraffin)
Unconjugated
DNMT3A
Liquid
DNMT3A
DNMT3A Fusion Protein Ag7015
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat