Learn More
DNMT3A Rabbit anti-Mouse, Unconjugated, Polyclonal, Active Motif™
Rabbit Polyclonal Antibody
Manufacturer:  Active MotifSupplier Diversity Partner 39897
Encompass
 View more versions of this product
Catalog No. 50-199-2940
Add to cart
Description
Description
Methylation of DNA at cytosine residues plays an important role in regulation of gene expression, genomic imprinting and is essential for mammalian development. Hypermethylation of CpG islands in tumor suppressor genes or hypomethylation of bulk genomic DNA may be linked with development of cancer. To date, 3 families of mammalian DNA methyltransferase genes have been identified which include Dnmt1, Dnmt2 and Dnmt3. Dnmt1 is constitutively expressed in proliferating cells and inactivation of this gene causes global demethylation of genomic DNA and embryonic lethality. Dnmt2 is expressed at low levels in adult tissues and its inactivation does not affect DNA methylation or maintenance of methylation. The Dnmt3 family members, Dnmt3a and Dnmt3b, are strongly expressed in ES cells but their expression is down regulated in differentiating ES cells and is low in adult somatic tissue. Recently, it has been shown that naturally occurring mutations of Dnmt3b gene occurs in patients with a rare autosomal recessive disorder, termed ICF (immunodeficiency, centromeric instability, and facial anomalies) syndrome.
Specifications
Specifications
DNMT3A
Western Blot
Unconjugated
DNMT3A
Liquid
Rabbit
IgG
100 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
13435
DNMT3A
Lot-Specific
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
O88508
AU015806; DNA (cytosine-5-)-methyltransferase 3 alpha; DNA (cytosine-5)-methyltransferase 3A; DNA cytosine methyltransferase 3A2; DNA methyltransferase 3 alpha; DNA methyltransferase 3A; DNA methyltransferase HsaIIIA; DNA methyltransferase MmuIIIA; DNA MTase HsaIIIA; DNA MTase MmuIIIA; Dnmt3a; DNMT3A2; M.HsaIIIA; MmuIIIA; TBRS; W91664
This DNMT3A antibody was raised against a recombinant protein corresponding to amino acids 1-247 of mouse DNMT3A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse