Learn More
Abnova™ DNMT3L 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00029947T01
Catalog No. 89-942-285
Add to cart
Specifications
Specifications
Western Blot
293T
BC002560.2
pCMV-DNMT3L full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
DNMT3L 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 42.57kDa
Transient overexpression cell lysate was tested with anti-DNMT3L antibody (Mfr. No. H00029947-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)