Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3L Mouse anti-Human, Mouse, Clone: S117-9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527628

Catalog No. PIMA527628


Add to cart

Description

Description

CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. This gene encodes a nuclear protein with similarity to DNA methyltransferases. This protein is not thought to function as a DNA methyltransferase as it does not contain the amino acid residues necessary for methyltransferase activity. However, this protein does stimulate de novo methylation by DNA cytosine methyltransferase 3 alpha and it is thought to be required for the establishment of maternal genomic imprints. This protein also mediates transcriptional repression through interaction with histone deacetylase 1. Alternative splicing results in two transcript variants. An additional splice variant has been described but its biological validity has not been determined.
Specifications

Specifications

DNMT3L
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Q9CWR8, Q9UJW3
Cytosine 5 methyltransferase 3 like protein, DNA (cytosine 5 ) methyltransferase 3 like, DNA (cytosine-5)-methyltransferase 3-like, DNA cytosine 5 methyltransferase 3 like protein, DNA methyltransferase 3 like protein, DNM3L_HUMAN, Dnmt 3L, Dnmt3l, Human cytosine 5 methyltransferase 3 like protein, MGC1090
Mouse
IgG1
100 μg
-20°C
Primary
29947, 54427
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
S117-9
Unconjugated
DNMT3L
Liquid
DNMT3L
Fusion protein amino acids 1-387 of human DNMT3L
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit