Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNPEP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100483
Description
Antibody detects endogenous levels of total DNPEP.
Aminopeptidase with specificity towards an acidic amino acid at the N-terminus. Likely to play an important role in intracellular protein and peptide metabolism.
Specifications
DNPEP | |
Polyclonal | |
Unconjugated | |
DNPEP | |
AA407814; Aspartyl aminopeptidase; ASPEP; DAP; Dnpep | |
Rabbit | |
Affinity chromatography | |
RUO | |
23549 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9ULA0 | |
DNPEP | |
A synthesized peptide derived from human DNPEP(Accession Q9ULA0), corresponding to amino acid residues I337-S387. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction