Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ dNTP Set Promotion

Optimize molecular biology applications with this dNTP set, consisting of aqueous solutions of dATP, dTTP, dCTP and dGTP supplied in separate tubes.

$130.50 - $405.00

Specifications

For Use With (Application) For use in all molecular biology applications including PCR, real-time PCR, high fidelity and long PCR, LAMP-PCR, cDNA synthesis, RT-PCR, RDA, MDA, DNA labeling and DNA sequencing.
Purity >99% (by HPLC)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FERR0181PM
View Documents
Thermo Scientific™
R0181
4 x 0.25mL (4 x 25 μmol of 100mM solution) Each for $130.50
Add to cart
 
FERR0182PM
View Documents
Thermo Scientific™
R0182
4 x 1mL (4 x 100 μmol of 100mM solution) Each for $405.00
Add to cart
 
Description

Description

dNTP Set is a convenient set of 100 mM aqueous solutions of each of dATP, dCTP, dGTP and dTTP, supplied in separate vials. The solutions of these extremely stable nucleotides are titrated to pH 7.3-7.5 with NaOH. The nucleotides have greater than 99% purity, are free of nuclease activities, human and E. coli DNA. They are designed for many different molecular biology applications.

Highlights

  • Greater than 99% purity confirmed by HPLC
  • Free ofhuman and E. coli DNA
  • Application tested in:
    • Long range PCR (40 kb)
    • cDNA synthesis and RT-PCR
    • Real-time PCR
    • Standard PCR
    • High fidelity PCR
  • Highly stable
    • Stable after multiple freeze-thaw cycles
    • Up to 95% of dNTPs remain in triphosphate form even after 7 weeks at room temperature
    • Up to 90% of dNTPs remain in triphosphate form after 30 cycles of PCR (1 min at 94°C; 3 min at 72°C)
Specifications

Specifications

For use in all molecular biology applications including PCR, real-time PCR, high fidelity and long PCR, LAMP-PCR, cDNA synthesis, RT-PCR, RDA, MDA, DNA labeling and DNA sequencing.
>99% (by HPLC)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit