Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNTT Mouse anti-Human, Unconjugated, Clone: TdT/1391, Abnova
Mouse Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB20916
Description
DNTT Monoclonal antibody specifically detects DNTT in Human samples. It is validated for ELISA, IHC-P.Specifications
DNTT | |
TdT/1391 | |
deoxynucleotidyltransferase, terminal | |
Liquid | |
Mouse | |
IgG1, Kappa | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
Monoclonal | |
Human |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
DNTT | |
TDT | |
Recombinant protein corresponding to amino acids 85-215 of human DNTT | |
100 μg | |
Primary | |
1791 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title