Learn More
DOCK10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 168341AP
Catalog No. 50-173-1210
Add to cart
Description
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Overexpression of this gene results in a decrease in the processing of insulin and hepatocyte growth factor receptors to their mature subunits. This decrease is caused by the mislocalization of furin, the endoprotease responsible for cleavage of insulin and hepatocyte growth factor receptors. This protein is involved in endosomal trafficking from the plasma membrane to recycling endosomes or the trans-Golgi network. This gene encodes two transcript variants encoding distinct isoforms.
Specifications
Specifications
DOCK10
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BZN6, Q96BY6
dedicator of cytokinesis 10, DOCK10, DRIP2, KIAA0694, Nbla10300, ZIZ3, Zizimin 3
Rabbit
IgG
150 μL
-20°C
Primary
210293, 301556, 55619
Immunoprecipitation
Unconjugated
DOCK10
Liquid
DOCK10
DOCK10 Fusion Protein Ag10486
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat