Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DOCK11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563767
Description
Immunogen sequence: KDTVETAQDD ETSSQGKAEN IMASLERSMH PELMKYGRET EQLNKLSRGD GRQNLFSFDS EVQRLDFSGI EPDIKPFEEK CNKRFLVNCH DLTF Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Small GTPases of the Rho family, Rho, Rac, and Cdc42, are critical regulators of the actin cytoskeleton and many other cellular processes. Rho GTPases are activated by Dbl-homology (DH)-domain-containing guanine nucleotide exchange factors (GEFs). DOCK 11 (dedicator of cytokinesis 11), also known as ACG or ZIZ2 (zizimin2), is a 2073 amino acid protein belonging to the DOCK family of cytokinesis-regulating proteins that is mainly expressed in peripheral blood leukocytes. DOCK 11 functions as a GEF that binds and activates Cdc42 by exchanging bound GDP for free GTP. Cdc42 mediates cell polarity, gene expression, cell cycle progression and cell-cell contacts. Similar to other DOCK family members, DOCK 11 contains a PH domain and two internal DOCK homology regions designated DHR1 and DHR2.
Specifications
DOCK11 | |
Polyclonal | |
Unconjugated | |
DOCK11 | |
5033414A21Rik; 8030476J24; ACG; activated Cdc42-associated guanine nucleotide exchange factor; Activated Cdc42-associated guanine nucleotide exchange factor, ACG; bB128O4.1; bB128O4.1.1 (novel protein); dedicator of cytokinesis 11; dedicator of cytokinesis protein 11; Dock11; DOCK-11; sph238; spleen specific 238 kDa protein with PH domain; ZIZ2; Zizimin 2; zizimin2; Zizimin-2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
139818 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JSL3 | |
DOCK11 | |
Recombinant protein corresponding to Human DOCK11. Recombinant protein control fragment (Product #RP-101934). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction