Learn More
DOCK180 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 202441AP
Catalog No. 50-173-2790
Add to cart
Description
Description
The antibody is specific to DOCK1.
This gene encodes a loosely associated peripheral membrane protein related to the LanC family of bacterial membrane-associated proteins involved in the biosynthesis of antimicrobial peptides. This protein may play a role as a peptide-modifying enzyme component in eukaryotic cells. Previously considered a member of the G-protein-coupled receptor superfamily, this protein is now in the LanC family. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
Specifications
DOCK180
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14185, Q8BUR4
ced5, dedicator of cytokinesis 1, DOCK1, DOCK180
Rabbit
IgG
150 μL
-20°C
Primary
1793, 309081, 330662
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
DOCK1
Liquid
Dock1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat