Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOCK2 Monoclonal Antibody (OTI4E12)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526548

Catalog No. PIMA526548


Only null left
Add to Cart

Description

Description

DOCK2 Monoclonal Antibody for IHC (P)

Dedicator of cytokinesis 2 (DOCK2) is a gene located on chromosome 5q15, encoding a member of the DOCK family of guanine nucleotide exchange factors (GEFs). DOCK2 plays a crucial role in the immune system by regulating the actin cytoskeleton, which is essential for cell movement and immune cell activation. It is predominantly expressed in hematopoietic cells, including lymphocytes and other leukocytes, where it facilitates chemotaxis and polarization by activating the small GTPase Rac. This activation is critical for processes such as lymphocyte homing, migration towards chemokines, and phagocytosis. Genetic mutations or dysregulation of DOCK2 can lead to immunodeficiency, characterized by impaired immune responses and increased susceptibility to infections. Research into DOCK2 focuses on its therapeutic potential in modulating immune responses and its role in disorders characterized by immune dysregulation, such as autoimmune diseases and chronic inflammation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DOCK2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q92608
DOCK2
Human recombinant protein fragment corresponding to amino acids 1544-1830 of DOCK2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin)
OTI4E12
Unconjugated
DOCK2
AI662014; AW122239; CED-5; dedicator of cytokinesis 2; dedicator of cyto-kinesis 2; dedicator of cytokinesis protein 2; DOCK2; FLJ46592; Hch; IMD40; KIAA0209; LOW QUALITY PROTEIN: dedicator of cytokinesis protein 2; MBC; Protein Hch
Mouse
Affinity chromatography
RUO
1794
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.