Learn More
DOCK8 Mouse anti-Human, Clone: OTI8D9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF801833
Catalog No. 50-166-5742
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Dedicator of cytokinesis protein 8 (DOCK8) is a member of the DOCK180 family of guanine nucleotide exchange factors. DOCK8 plays an essential role in humoral immune responses and is important in the proper formation of the B cell immunological synapse. Mutations in this gene result in the autosomal recessive form of the hyper-IgE syndrome.
Specifications
Specifications
DOCK8
Monoclonal
Unconjugated
DOCK8
Lyophilized
DOCK8
Human recombit protein fragment corresponding to amino acids 84-405 of human DOCK8 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI8D9
PBS with 8% trehalose and no preservative; pH 7.3
Q8NF50
HEL-205, MRD2, ZIR8
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
81704