Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK9 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500264AF594
Description
DOCK9 Polyclonal antibody specifically detects DOCK9 in Human samples. It is validated for Western BlotSpecifications
DOCK9 | |
Polyclonal | |
Western Blot | |
Cdc42 guanine nucleotide exchange factor zizimin-1, dedicator of cytokinesis 9, dedicator of cytokinesis protein 9, DKFZp686C11110, DKFZp686D2047, DKFZp686N04132, FLJ11949, FLJ16744, FLJ45601, KIAA1058FLJ44528, KIAA1085, ZIZ1FLJ45282, ZIZIMIN1 | |
The immunogen recognized by this antibody maps to a region between residues 1250 and 1300 of human Dedicator of Cytokinesis 9 using the numbering given in entry NP_056111.1 (GeneID 23348). | |
0.1 mL | |
Apoptosis, Cell Biology, Signal Transduction | |
23348 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction