Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Docusate Sodium, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
577-11-7
C20H37NaO7S
444.56
APSBXTVYXVQYAB-UHFFFAOYNA-M
sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
577-11-7
C20H37NaO7S
444.56
APSBXTVYXVQYAB-UHFFFAOYNA-M
sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

DO105, 577-11-7, C20H37NaO7S

Spectrum™ Docusate Sodium, USP is used to treat occasional constipation. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Ignition Residue 15.5-16.5%
Packaging Amber Glass Bottle
Percent Purity 99 to 100.5%
Quantity 125 g