Alfa Aesar™ Dodecacarbonyltetracobalt, 98% min

Manufacturer: Alfa Aesar™ 1305503

Quantity 1g

 View more versions of this product

Catalog No. AA1305503

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 17786-31-1
    Linear Formula Co4(CO)12
    Formula Weight 571.86
    MDL Number MFCD00064698
    UN Number UN3179
    Synonym Tetracobalt dodecacarbonyl
    Chemical Name or Material Dodecacarbonyltetracobalt

    Specifications

    Formula Weight 571.86
    Health Hazard 3 GHS P Statement
    P210-P280h-P262-P304+P340-P309-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    RTECS Number GG3410000
    UN Number UN3179
    Health Hazard 1 Danger
    Melting Point 60°C (dec.)
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
    Density 2.09g/mL
    MDL Number MFCD00064698
    Color Black
    Physical Form Crystalline
    Linear Formula Co4(CO)12
    CAS 17786-31-1
    Synonym Tetracobalt dodecacarbonyl
    Health Hazard 2 GHS H Statement
    H228-H301-H331-H351-H317
    Flammable solid.
    Toxic if swallowed.
    Toxic if inhaled.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Sensitivity Air Sensitive
    Quantity 1g
    Chemical Name or Material Dodecacarbonyltetracobalt
    Assay ≤98%

    GHS H Statement
    H228-H301-H331-H351-H317
    Flammable solid.
    Toxic if swallowed.
    Toxic if inhaled.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P280h-P262-P304+P340-P309-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit